Learn More
Selleck Chemical LLC Metoclopramide HCl S4289-50mg
SDP
Supplier:  Selleck Chemical LLC S428950MG
Encompass_Preferred
Metoclopramide HCl(Metoclopramide hydrochloride) is a selective dopamine D2 receptor antagonist used for the treatment of nausea and vomiting
Catalog No. 50-310-9435