Learn More
Selleck Chemical LLC Metoclopramide HCl S4289-200mg
SDP
Supplier:  Selleck Chemical LLC S4289200MG
Encompass_Preferred
Metoclopramide HCl(Metoclopramide hydrochloride) is a selective dopamine D2 receptor antagonist used for the treatment of nausea and vomiting
Catalog No. 50-311-2184