Learn More
Medchemexpress LLC Metolachlor-d6 | 1219803-97-0 | 99.56% | 5 MG
SDP
Supplier:  Medchemexpress LLC HYB1871S5MG
Encompass_Preferred
Metolachlor-d6 is the deuterium labeled Metolachlor, a pre-emergent selective, chloroacetanilide herbicide. It is used to control various annual grass and broadleaf weeds in corn and other crops, and is a chiral herbicide composed of four stereoisomers. Deuterium substitution can influence the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-004-12144