Learn More
Medchemexpress LLC Metolazone-d7 | 2714484-71-4 | 99.90% | 1 MG
SDP
Supplier:  Medchemexpress LLC HYB0209S1MG
Encompass_Preferred
Metolazone-d7 is deuterium labeled Metolazone, primarily used to treat congestive heart failure and high blood pressure. This compound can be used as a tracer and as an internal standard for quantitative analysis. Deuteration has gained attention because of its potential to affect the pharmacokinetic and metabolic profiles of drugs.
  • Used as a tracer
  • Functions as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Deuterium substitution impacts pharmacokinetics
Catalog No. 50-004-05399