Learn More
Medchemexpress LLC HY-129441 10mg Medchemexpress, Metoprine CAS:7761-45-7 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY12944110MG
Encompass_Preferred
Medchemexpress, HY-129441 10mg Metoprine CAS:7761-45-7 Metoprine (BW 197U) is a potent histamine N-methyltransferase (HMT) inhibitor. Metoprine, a diaminopyrimidine derivative, can cross the blood-brain barrier and increase brain histamine levels by inhibiting HMT[2]. Metoprine is an antifolate and antitumor agent[3]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-203-0156