Learn More
Selleck Chemical LLC Metoprolol 25mg 51384-51-1
SDP
Supplier:  Selleck Chemical LLC S543025MG
Encompass_Preferred
Metoprolol is a cardioselective ?1-adrenergic blocking agent with log Kd values of ?7.26¿0.07, ?6.89¿0.09, ?5.16¿0.12 for ?1, ?2, and ?3 adrenoceptors, respectively. It is used for acute myocardial infarction, heart failure, angina pectoris and mild to moderate hypertension. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-194-8430