Learn More
Selleck Chemical LLC Metoprolol S5430-1g
SDP
Supplier:  Selleck Chemical LLC S54301G
Encompass_Preferred
Metoprolol is a cardioselective 1-adrenergic blocking agent with log Kd values of 7 26 0 07 6 89 0 09 5 16 0 12 for 1 2 and 3 adrenoceptors respectively It is used for acute myocardial infarction heart failure angina pectoris and mild to moderate hypertension
Catalog No. 50-310-9954