Learn More
Medchemexpress LLC Metoprolol | 51384-51-1 | MFCD00599534 | 99.99% | 267.36 | 500 MG
SDP
Supplier:  Medchemexpress LLC HY17503500MG
Encompass_Preferred
Metoprolol is an orally active, selective β1-adrenoceptor antagonist. It also demonstrates anti-inflammation, antitumor, and anti-angiogenic properties.
  • Orally active
  • Selective β1-adrenoceptor antagonist
  • Shows anti-inflammation properties
  • Shows antitumor properties
  • Shows anti-angiogenic properties
Catalog No. 50-003-86767