Learn More
Medchemexpress LLC Metoprolol-d7 hydrochloride | 1219798-61-4 | 99.7% | 310.87 | 1 MG
SDP
Supplier:  Medchemexpress LLC HY17503S1MG
Encompass_Preferred
Metoprolol-d7 (hydrochloride) is the deuterium labeled Metoprolol. Metoprolol is an orally active, selective β1-adrenoceptor antagonist, demonstrating anti-inflammatory, antitumor, and anti-angiogenic properties. It is ideal for use as a tracer and as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Used as a tracer
  • Functions as an internal standard for quantitative analysis
  • Shows anti-inflammation, antitumor, and anti-angiogenic properties
Catalog No. 50-003-86799