Learn More
Medchemexpress LLC Metoprolol-d7 (hydrochloride) | 1219798-61-4 | 99.7% | 310.87 g·mol⁻1 | C15H19D7ClNO3 | 10 MG
SDP
Supplier:  Medchemexpress LLC HY17503AS10MG
Encompass_Preferred
Metoprolol-d7 (hydrochloride) is the deuterium-labeled hydrochloride salt of metoprolol supplied as a high-purity stable isotope standard for quantitative bioanalysis. It is intended for use as an internal standard in mass spectrometry assays to improve accuracy and precision in pharmacokinetic and bioanalytical studies.
  • High isotopic purity for accurate quantitation.
  • High chemical purity (99.7%) to minimize interference.
  • Hydrochloride salt form with molecular weight 310.87 g·mol⁻1.
  • Supplied in small milligram quantities suitable for analytical standards.
  • Water-soluble with ultrasonic warming recommended for dissolution.
Catalog No. 50-002-99134