Learn More
Medchemexpress LLC Metoprolol (Standard) | 51384-51-1 | MFCD00868158 | 99.98% | 267.36 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY17503R50MG
Encompass_Preferred
Metoprolol (Standard) is the analytical standard of Metoprolol, intended for research and analytical applications. It is an orally active, selective β1-adrenoceptor antagonist. Metoprolol exhibits anti-inflammation, antitumor, and anti-angiogenic properties. It is commonly used in qualitative, quantitative, and methodological research experiments in techniques such as HPLC, GC, and MS.
  • Is an analytical standard.
  • Is intended for research and analytical applications.
  • Is an orally active compound.
  • Functions as a selective β1-adrenoceptor antagonist.
  • Shows anti-inflammation properties.
  • Shows antitumor properties.
  • Shows anti-angiogenic properties.
  • Is suitable for use in HPLC, GC, and MS.
Catalog No. 50-003-86803