Learn More
Medchemexpress LLC Metoprolol succinate | 98418-47-4 | 99.6% | 500 MG
SDP
Supplier:  Medchemexpress LLC HY17503A500MG
Encompass_Preferred
Metoprolol succinate is an orally active, selective β1-adrenoceptor antagonist. It exhibits anti-inflammatory, antitumor, and anti-angiogenic properties.
  • Selective β1-adrenoceptor antagonist.
  • Shows cytotoxic effects on U937 and MOLT-4 cells.
  • Reduces proinflammatory cytokines and atherosclerosis in mice.
  • Exhibits anti-inflammatory and anti-viral effects in a murine model of viral myocarditis.
  • Decreases activated caspase-9 protein expression and inhibits myocardial apoptosis in rats.
Catalog No. 50-003-87005