Learn More
Medchemexpress LLC Metoprolol succinate | 98418-47-4 | 99.6% | 652.82 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY17503A100MG
Encompass_Preferred
Metoprolol succinate is an orally active, selective β1-adrenoceptor antagonist. It exhibits anti-inflammatory, antitumor, and anti-angiogenic properties. Metoprolol has also shown cytotoxic effects on U937 and MOLT-4 cells in a dose and time-dependent manner.
  • Orally active
  • Selective β1-adrenoceptor antagonist
  • Anti-inflammation properties
  • Antitumor properties
  • Anti-angiogenic properties
Catalog No. 50-003-86800