Learn More
Medchemexpress LLC Metoprolol succinate | 98418-47-4 | 99.6% | 652.82 | 200 MG
SDP
Supplier:  Medchemexpress LLC HY17503A200MG
Encompass_Preferred
Metoprolol succinate is an orally active, selective β1-adrenoceptor antagonist. It shows anti-inflammation, antitumor and anti-angiogenic properties.
  • Exhibits cytotoxic effects on U937 and MOLT-4 cells.
  • Reduces proinflammatory cytokines and atherosclerosis in ApoE-/- Mice.
  • Demonstrates anti-inflammation and anti-virus effects in murine model with coxsackievirus B3-induced viral myocarditis.
  • Decreases activated caspase-9 protein expression.
  • Inhibits myocardial apoptosis.
Catalog No. 50-003-87185