Learn More
Medchemexpress LLC Metoprolol (succinate) (standard) | 98418-47-4 | 99.9% | 652.82 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY17503AR
Encompass_Preferred
Metoprolol succinate (Standard) is the analytical standard of Metoprolol succinate, intended for research and analytical applications. It is an orally active, selective β1-adrenoceptor antagonist that exhibits anti-inflammation, antitumor, and anti-angiogenic properties.
  • Used in qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS
  • Orally active, selective β1-adrenoceptor antagonist
  • Exhibits anti-inflammation, antitumor, and anti-angiogenic properties
Catalog No. 50-003-84116