Learn More
Medchemexpress LLC Metoprolol succinate (Standard) | 98418-47-4 | 99.94% | 652.82 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY17503AR50MG
Encompass_Preferred
Metoprolol (succinate) (Standard) is the analytical standard of Metoprolol (succinate). This product is intended for research and analytical applications. Metoprolol succinate is an orally active, selective β1-adrenoceptor antagonist and shows anti-inflammation, antitumor and anti-angiogenic properties. It is commonly used in qualitative, quantitative and methodological research experiments in HPLC, GC and MS.
  • Analytical standard
  • Orally active, selective β1-adrenoceptor antagonist
  • Anti-inflammation, antitumor and anti-angiogenic properties
  • Appearance: Solid
  • Color: White to off-white
Catalog No. 50-003-86863