Learn More
Medchemexpress LLC Metoprolol (succinate) (standard) | 98418-47-4 | MFCD23115896 | 99.8% | 652.82 | C34H56N2O10 | 10 MG

Supplier: Medchemexpress LLC HY17503AR10MG
Metoprolol succinate (standard) is an analytical reference standard of the succinate salt of metoprolol, a selective β1-adrenoceptor antagonist. It is intended for research and analytical use, including method development, assay validation, and quality control in laboratory settings.
- High purity suitable for analytical use (assay 99.82%).
- Provided as a stable solid in amber/brown glass packaging.
- Molecular formula C34H56N2O10 and molecular weight 652.82.
- CAS number 98418-47-4 for unambiguous identification.
- Suitable for method development, assay calibration, and quality control.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.