Learn More
Apexbio Technology LLC Metoprolol Tartrate 56392-17-7 10mM (in 1mL DMSO)
SDP
Supplier:  Apexbio Technology LLC B13395.1
Encompass_Preferred
Metoprolol Tartrate (CAS 56392-17-7) is a selective 1-adrenergic receptor antagonist employed primarily for cardiovascular research purposes By specifically inhibiting 1-adrenergic receptors located predominantly in cardiac tissue it reduces sympathetic influences on heart rate and myocardial contractility Clinical research has indicated that genetic polymorphisms in 1-adrenergic receptors significantly affect individual hypertensive responses to treatment with metoprolol Due to its targeted mode of action and known genetic modulatory effects it is widely utilized in hypertension and cardiovascular pharmacogenomics studies
Catalog No. 50-000-62065