Learn More
Medchemexpress LLC Metronidazole | 443-48-1 | 99.9% | 1 ML
SDP
Supplier:  Medchemexpress LLC HYB031810MM/1ML
Encompass_Preferred
Metronidazole is an orally active nitroimidazole antibiotic with broad activity against anaerobic bacteria and protozoa, known for its ability to cross the blood-brain barrier. It is effective in inhibiting various anaerobic protozoa and exhibits bactericidal activity against anaerobic bacteria. It also induces apoptosis in Blastocystis sp. cells. This product is provided as a 10 mM solution in 1 mL of DMSO for research into anaerobic infections.
  • Orally active nitroimidazole antibiotic.
  • Can cross the blood-brain barrier.
  • Effective against anaerobic protozoa, including Trichomonas vaginalis, Entamoeba histolytica, Giardia lamblia, and Balantidium coli.
  • Demonstrates bactericidal activity against anaerobic bacteria.
  • Induces apoptosis in Blastocystis sp. cells.
  • Provided as a 10 mM solution in DMSO.
Catalog No. 50-004-06691