Learn More
Medchemexpress LLC Metronidazole | 443-48-1 | 99.96% | 500 MG
SDP
Supplier:  Medchemexpress LLC HYB0318500MG
Encompass_Preferred
Metronidazole is an orally active nitroimidazole antibiotic that can cross the blood-brain barrier. It is used for research into anaerobic infections.
  • Inhibits anaerobic protozoa, such as Trichomonas vaginalis, Entamoeba histolytica, Giardia lamblia, and Balantidium coli.
  • Exhibits bactericidal activity against anaerobic bacteria.
  • Induces granular formation and apoptosis in Blastocystis sp.
Catalog No. 50-004-06711