Learn More
Selleck Chemical LLC Metronidazole 50mg 443-48-1
SDP
Supplier:  Selleck Chemical LLC S190750MG
Encompass_Preferred
Metronidazole, a synthetic antibacterial and antiprotozoal agent of the nitroimidazole class, is used against protozoa. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. NC1338211