Learn More
Medchemexpress LLC Metronidazole benzoate | 13182-89-3 | 99.9% | 100 MG

Supplier: Medchemexpress LLC HY122975100MG
Metronidazole benzoate is the benzoate ester of metronidazole supplied for research and analytical use. It acts as an antibacterial, antimicrobial, antiparasitic, and antitrichomonal agent and has reported in vitro cytotoxicity in several human cell lines.
- High purity: 99.9%.
- Molecular formula C13H13N3O4 and molecular weight 275.26 g/mol.
- Soluble in DMSO (250 mg/mL) and suitable for formulation in common in vivo vehicles.
- Available in multiple pack sizes, including 100 mg.
- Powder storage recommended at -20°C for long-term and 4°C for shorter term.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.