Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Metronidazole benzoate | 13182-89-3 | 99.9% | 500 MG
SDP

Supplier:  Medchemexpress LLC HY122975500MG

Encompass_Preferred

Metronidazole benzoate is the benzoyl ester of metronidazole used as an antibacterial, antimicrobial, antiparasitic, and antitrichomonal research reagent. It is provided as a high-purity analytical standard for studies of anaerobic and protozoan infections.

  • High purity (99.9%) analytical standard.
  • Suitable for research into anaerobic and protozoan infections.
  • Available in multiple pack sizes, including 500 MG.
  • Chemical formula C13H13N3O4 and molecular weight 275.26 g/mol.
  • Soluble in DMSO for preparation of stock solutions.

Catalog No. 50-002-30501


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.