Learn More
Medchemexpress LLC Metronidazole impurity 1 | 5006-68-8 | MFCD00159670 | 95.0% | 157.13 g/mol | C5H7N3O3 | 1 G
SDP
Supplier:  Medchemexpress LLC HYW0155091G
Encompass_Preferred
Metronidazole impurity 1 (2-(5-nitro-1H-imidazol-1-yl)ethanol; CAS 5006-68-8) is a characterized impurity used as an analytical reference standard for identification and quantification of metronidazole-related substances in research and quality control. Supplied for laboratory use only.
  • Used as an analytical reference standard for impurity identification and quantification.
  • Suitable for method development and validation in HPLC and GC analyses.
  • Characterized molecular weight 157.13 g/mol and formula C5H7N3O3.
  • Typical purity around 95% as reported by suppliers.
  • Intended for research and analytical use only; not for human consumption.
Catalog No. 50-003-02552