Learn More
Cayman Chemical MetronIdazole ImpurIty G 25mg
SDP
Supplier:  Cayman Chemical 4229125 MG
Encompass_Preferred
A potential impurity found in commercial preparations of metronidazole; an inactive metabolite of metronidazole
Catalog No. 50-294-2966