Learn More
Cayman Chemical MetronIdazole ImpurIty G 50mg
SDP
Supplier:  Cayman Chemical 4229150 MG
Encompass_Preferred
A potential impurity found in commercial preparations of metronidazole; an inactive metabolite of metronidazole
Catalog No. 50-294-3420