Learn More
Medchemexpress LLC Metronidazole (Standard) | 443-48-1 | 99.99% | 100 MG

Supplier: Medchemexpress LLC HYB0318R100MG
Metronidazole (Standard) is the analytical standard of Metronidazole. It is intended for research and analytical applications. Metronidazole is an orally active nitroimidazole antibiotic that can cross the blood-brain barrier and is used for the research of anaerobic infections.
- Analytical standard for metronidazole.
- Intended for research and analytical applications.
- Orally active nitroimidazole antibiotic.
- Can cross the blood-brain barrier.
- Used for the research of anaerobic infections.
- Commonly employed in qualitative, quantitative, and methodological research experiments such as HPLC, GC, and MS.
- Purity: 99.99%.
- Appearance: White to off-white solid.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.