Learn More
Sino Biological Oligo Substrates: METTL3/METTL14 Substrate
Supplier:  Sino Biological M32358025
Encompass_Preferred
The synthetic single-stranded 27 base’s oligo ribo-nucleotide is used as a substrate for assay of methyltransferase METTL3/METTL14 complex.
Catalog No. 50-256-8640