Learn More
Medchemexpress LLC HY-W015007 25mg Medchemexpress, Metyrosine CAS:672-87-7 Purity:>98%
Supplier: Medchemexpress LLC HYW01500725MG
Medchemexpress, HY-W015007 25mg Metyrosine CAS:672-87-7 Metyrosine is a selective tyrosine hydroxylase enzyme inhibitor. Metyrosine exerts anti-inflammatory and anti-ulcerative effects. Metyrosine significantly inhibits high COX-2 activity. Metyrosine is a very effective agent for blood pressure control. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.