Learn More
Medchemexpress LLC HY-113071A 5mg Medchemexpress, Mevalonic acid (lithium salt) CAS: Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY113071A5MG
Encompass_Preferred
Medchemexpress, HY-113071A 5mg Mevalonic acid (lithium salt) CAS: Mevalonic acid lithium salt, a precursor in the mevalonate pathway, is essential for cell growth and proliferation. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9223