Learn More
Selleck Chemical LLC Mevastatin S4223-1g
SDP
Supplier:  Selleck Chemical LLC S42231G
Encompass_Preferred
Mevastatin (ML-236B Compactin) is a competitive inhibitor of HMG-Coenzyme A (HMG-CoA) reductase with a binding affinity 10 000 times greater than the HMG-CoA substrate itself
Catalog No. 50-310-8701