Learn More
TARGETMOL CHEMICALS INC Mevastatin 500MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T0683500MG
Encompass_Preferred
Also available in 1 mL, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. Mevastatin (ML236B) is an HMG-CoA reductase inhibitor that was initially isolated from the mold Pythium ultimum. Mevastatin was the first statin to enter clinical trials. Purity 97.96%
Catalog No. 50-288-0523