Learn More
Medchemexpress LLC Mevastatin | 73573-88-3 | 390.51 | 1 ML
SDP
Supplier:  Medchemexpress LLC HY1740810MM/1ML
Encompass_Preferred
Mevastatin (Compactin) is the first HMG-CoA reductase inhibitor belonging to the statins class. It acts as a lipid-lowering agent, induces apoptosis, and arrests cancer cells in the G0/G1 phase. It also increases endothelial nitric oxide synthase (eNOS) mRNA and protein levels. This compound exhibits antitumor activity and shows potential for treating cardiovascular diseases.
  • HMG-CoA reductase inhibitor
  • Lipid-lowering agent
  • Induces apoptosis and arrests cancer cells in G0/G1 phase
  • Increases endothelial nitric oxide synthase (eNOS) mRNA and protein levels
  • Exhibits antitumor activity
  • Potential for cardiovascular diseases treatment
  • Targets metabolic enzyme/protease, autophagy, apoptosis, and anti-infection pathways
  • Research areas include infection, cardiovascular disease, neurological disease, metabolic disease, and cancer
Catalog No. 50-003-83986