Learn More
Abcam Mevastatin, HMG-CoA reductase inhibitor, 10MG
Supplier:  Abcam AB12065210MG
Encompass_Preferred
MW 390.5 Da, Purity >98%. Potent HMG-CoA reductase inhibitor. Proapoptotic, able to arrest cells in G₁ by down-regulation of CDK4, CDK6 and cyclin D1 and up-regulation of p21 and p27.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-7385