Learn More
Medchemexpress LLC HY-129395 10mg , Mezigdomide CAS:2259648-80-9 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY12939510MG
Encompass_Preferred
HY-129395 10mg Mezigdomide CAS: 2259648-80-9 Mezigdomide (CC-92480), a cereblon E3 ubiquitin ligase modulating drug (CELMoD), acts as a molecular glue. Mezigdomide shows high affinity to cereblon, resulting in potent antimyeloma activity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-225-9031