Learn More
Medchemexpress LLC Mezigdomide | 2259648-80-9 | 99.16% | 50 MG
SDP
Supplier:  Medchemexpress LLC HY12939550MG
Encompass_Preferred
Mezigdomide (CC-92480) is a cereblon E3 ubiquitin ligase modulating agent (CELMoD) that functions as a molecular glue. It exhibits high affinity to cereblon, leading to potent antimyeloma activity. It induces loss of Aiolos and Ikaros in PBMCs, activating T cells and increasing IL-2 and IFN-γ production.
  • Functions as a molecular glue
  • Exhibits high affinity to cereblon
  • Potent antimyeloma activity
  • Activates T cells and increases IL-2 and IFN-γ production
  • Effective in resistant cell lines
  • Induces apoptosis as a single agent
  • Shows significant synergy with NSC 34521
Catalog No. 50-003-38372