Learn More
Medchemexpress LLC MF-095 | 2241021-89-4 | 98.7% | 493.62 | 1 MG
SDP
Supplier:  Medchemexpress LLC HY1389781MG
Encompass_Preferred
MF-095 is an inhibitor of USP30. It promotes mitochondrial autophagy and can be utilized in research related to neurological diseases.
  • It is a USP30 inhibitor.
  • It promotes mitochondrial autophagy.
  • It can be used in neurological disease-related research.
Catalog No. 50-003-51287