Learn More
Medchemexpress LLC Mf-dh-300 | 2673403-86-4 | 99.5% | 330.38 | 50 MG
Supplier: Medchemexpress LLC 50MGHY176803
MF-DH-300 is a 15-PGDH inhibitor that can be used for the research of muscle disorders such as spinal muscular atrophy (SMA). For research use only. 15-PGDH (15-Hydroxyprostaglandin dehydrogenase) acts as a tumor suppressor in various organs (breast, colon, liver, lung, pancreas), with decreased expression linked to increased tumorigenesis. It metabolizes intracellular PGE2, preventing its binding to EP receptors and thereby suppressing PGE2 signaling.
- Applicable to the research of muscle disorders such as spinal muscular atrophy (SMA)
- Inhibits 15-PGDH (15-Hydroxyprostaglandin dehydrogenase)
- Metabolizes intracellular PGE2
- Suppresses PGE2 signaling