Learn More
AVANTOR MASTERFLEX (VWR LLC) MF PP 1 1/2 X 3/8 TRI BRB 6/PK
Supplier:  AVANTOR MASTERFLEX (VWR LLC) MFLX3120045
Encompass
Masterflex Fitting, Polypropylene, Straight, Sanitary Clamp to Hose Barb Adapter, 1 1/2IN Tri-Clamp x 3/8IN ID; 6/PK, Avantor
Catalog No. 50-274-2618