Learn More
Narishige International Usa Inc Narishige MF2 Microforge, Precision Cutting, Built-in Microscope, Digital Heater Display
Supplier:  Narishige International Usa Inc MF2
Encompass
The Narishige MF2 Microforge is a versatile instrument designed for precisely crafting micropipette tips and glass microtools. It combines cutting, bending, and polishing functionalities with built-in microscope visualization, enhancing operational precision. Features like a stabilized XY control unit, a digital heater display, and user-friendly enhancements ensure high-quality tip finishing, making it essential for demanding applications such as patch clamp experiments.
Catalog No. NC2167623
Only null left
Add to Cart
Request a Quote