Learn More
AdipoGen MFAP4 human pAb
SDP
Supplier:  AdipoGen AG25A0061C100
Encompass_Preferred
Polyclonal Antibody. Recognizes human MFAP4. Detects a band of ~36kDa by Western blot. Source Rabbit. Applications ELISA, WB. Liquid. 0.2µm-filtered solution in PBS, pH 7.4. Contains no preservatives. Microfibril-associated protein 4 MFAP4 was initially identified as a gene commonly deleted in the contiguous Smith-Magenis syndrome SMS gene. MFAP4 was identified as growth factor to support expansion of hematopoietic stem cells in culture. It is involved in calcium-dependent cell adhesion and intercellular interactions.
Catalog No. 50-256-3251