Learn More
American Radiolabeled Chemicals Inc MFZ 2-12-3H 50UC
SDP
Supplier:  American Radiolabeled Chemicals Inc ART 119850UC
Encompass_Preferred
MFZ 2-12-3H 50uCi in ethanol, 1 mCi/ml, S.A. 50-60 Ci/mmol, M.W. 328.2, Shipped in Dry Ice, Exclusive, (2-beta-Carbomethoxy-3-beta-(3,4-dichlorophenyl) tropane)
Catalog No. 50-901-04879