Learn More
Medchemexpress LLC HY-18964 10mg , MG-101 CAS:110044-82-1 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1896410MG
Encompass_Preferred
HY-18964 10mg MG-101 CAS: 110044-82-1 MG-101 is a potent inhibitor of cysteine proteases which inhibits calpain I, calpain II, cathepsin B and cathepsin L with Kis of 190, 220, 150 and 500 pM, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-225-9784