Learn More
Selleck Chemical LLC MG-101
SDP
Supplier:  Selleck Chemical LLC S7386100MG
Encompass_Preferred
MG-101 (ALLN Calpain inhibitor-1 Ac-LLnL-CHO) is a cell-permeable and potent inhibitor of cysteine proteases including calpains and lysosomal cathepsins
Catalog No. 50-000-00087