Learn More
Medchemexpress LLC MG-115 | 133407-86-0 | 99.6% | 461.59 | 10 MG
SDP
Supplier:  Medchemexpress LLC HY1085521MG
Encompass_Preferred
MG-115 is a potent and reversible proteasome inhibitor. It has Kis of 21 nM and 35 nM for 20S and 26S proteasome, respectively. It specifically inhibits the chymotrypsin-like activity of the proteasome and induces p53-dependent apoptosis.
  • Potent and reversible proteasome inhibitor.
  • Specifically inhibits the chymotrypsin-like activity of the proteasome.
  • Induces p53-dependent apoptosis.
  • High purity (99.61%).
  • Intended for research use only.
Catalog No. 50-003-46233