Learn More
Sigma Aldrich Fine Chemicals Biosciences MG-132, Ready Made Solution, 200 ul
Supplier:  Sigma Aldrich Fine Chemicals Biosciences M7449200UL
Encompass
≥90% (HPLC). MG-132, Ready Made Solution is supplied as a 10 mM (1 mg/210 μL) 0.2 μm-filtered solution in DMSO. MG-132, Ready Made Solution has been used to inhibit proteasome.
Catalog No. NC1599764