Learn More
Apexbio Technology LLC MG-132, 500mg. Cas: 133407-82-6 MFCD: MFCD00674886. Proteasome inhibitor, Cell permeable, reversible
SDP
Supplier:  Apexbio Technology LLC A2585500
Encompass_Preferred
MG132 (carbobenzoxy-Leu-Leu-leucinal) as a peptide aldehyde effectively blocks the proteolytic activity of proteasome complex.9 Proteasome inhibitors including MG132 have been shown to induce apoptotic cell death through formation of ROS. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-4749
 Restricted Item