Learn More
Selleck Chemical LLC MG-132 5mg 1211877-36-9
SDP
Supplier:  Selleck Chemical LLC S26195MG
Encompass_Preferred
MG132 is a potent cell-permeable proteasome and calpain inhibitor with IC50s of 0.1 ?M and 1.2 ?M for the inhibition of proteasome and calpain, respectively. MG132 activates autophagy and induces apoptosis in tumor cells. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-833-8