Learn More
Medchemexpress LLC MG-2119 | 1065500-40-4 | 99.68% | 487.55 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY13930750MG
Encompass_Preferred
MG-2119 is a potent monomeric tau and α-syn aggregation inhibitor, investigated as a potential agent for neurological disorders research. For optimal stability, store this product at 4°C, sealed, away from moisture and light. In solvent, store at -80°C for up to 6 months or -20°C for 1 month under similar sealed conditions.
  • Potent monomeric tau and α-syn aggregation inhibitor
  • Potential agent for neurological disorders research
  • Store at 4°C sealed, away from moisture and light
Catalog No. 50-003-51351