Learn More
Ubpbio MG132, 100MG
Supplier:  Ubpbio F1102
Encompass_Preferred
Inhibitors – MG-132 (UBPBio catalog# F1100, F1101, F1102) – MG-132 is a reversible proteasome inhibitor that belongs to the family known as peptide aldehydes. MG-132 is cell permeable and can be used to treat various mammalian cells to block proteasome activities. MG-132 can be dissolved in DMSO for stock solution up to 20 mM. The typical concentrations for cell culture use are 1-10 µM. For in vitro use, the typical concentrations are 50-100 µM. CAS#: 133407-82-6
Catalog No. 50-112-6891